Description
- Closure type snap ring seal
- For snap ring vials
Specifications
Specifications
| Material (Liner) | PTFE |
| Diameter (Metric) Opening | 11 mm |
| Material | PTFE |
| Pre-slit | No |
| Material (Seal) | Polypropylene |
| No. per Pack | 100 Pk. |
| For Use With (Equipment) | Wide Mouth Crimp/Snap Seal Vials |