Learn More
Chem Service Inc Clothianidin, 210880-92-5, MFCD06200753, 100 mg
SDP
Supplier:  Chem Service Inc N11493100MG
Encompass_Preferred
Molecular Formula: C6-H8-Cl-N5-O2-S, MW: 249.68, 99.5%.
Catalog No. NC9087882
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.