Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cloxacillin Sodium, G-Biosciences
SCI_ED

Catalog No. S973278
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S973278 5 g
S000032 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S973278 Supplier G-Biosciences Supplier No. RC983
Add to Cart
Add to Cart

Cloxacillin sodium is a sodium salt formed from a chlorinated semisynthetic derivative of penicillin.

It has been shown to exhibit significant bactericidal activity against penicillin G-resistant staphylococci.

Chemical Identifiers

Molecular Weight (g/mol) 475.88 g/mol

Specifications

Boiling Point 689°C
Quantity 5 g
Percent Purity >98%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.