Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Clozapine, Atypical Antipsychotic, Solid, 326.8 Da, 50 mg

Catalog No. 502707930
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7930 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7930 Supplier Abcam Supplier No. AB12001950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Atypical antipsychotic compound with broad receptor antagonism and muscarinic agonist activity.

  • Molecular Weight: 326.8 Da
  • Purity: >98%
  • Form: Solid
  • Activity: Atypical antipsychotic compound; displays broad antagonism at D4, D2, 5-HT2A, 5-HT2C, 5-HT3, 5-HT6, and 5-HT7 receptors, and acts as an agonist at muscarinic receptors
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule D4, D2, 5-HT2A, 5-HT2C, 5-HT3, 5-HT6, 5-HT7, Muscarinic Receptors
Shipping Condition Ambient
Content And Storage Store at 4°C.
Form Solid
Quantity 50 mg
For Use With (Application) Atypical antipsychotic research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.