Learn More
Abcam Clozapine, Atypical Antipsychotic, Solid, 500 mg
Description
- Compound type: Atypical antipsychotic
- Purity: >98%
- Molecular Weight: 326.8 Da
- Biological activity: Antagonist at D4, D2, 5-HT2A, 5-HT2C, 5-HT3, 5-HT6, and 5-HT7 receptors; agonist at muscarinic receptors
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | D4 receptor, D2 receptor, 5-HT2A receptor, 5-HT2C receptor, 5-HT3 receptor, 5-HT6 receptor, 5-HT7 receptor, Muscarinic receptor |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Quantity | 500 mg |
| For Use With (Application) | Antipsychotic research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.