Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clozapine, MP Biomedicals

Catalog No. ICN15976005 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 mg

Catalog No. ICN15976005

Supplier: MP Biomedicals, Inc 0215976005

Chemical Identifiers

CAS 5786-21-0
Molecular Formula C18H19ClN4
Molecular Weight (g/mol) 326.828
InChI Key ZUXABONWMNSFBN-UHFFFAOYSA-N
Synonym clozapine, leponex, clozapin, clozaril, fazaclo, clorazil, iprox, clozapinum, asaleptin, clozapina
PubChem CID 2818
IUPAC Name 3-chloro-6-(4-methylpiperazin-1-yl)-5H-benzo[b][1,4]benzodiazepine
SMILES CN1CCN(CC1)C2=C3C=CC=CC3=NC4=C(N2)C=C(C=C4)Cl

Specifications

Quantity 5 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.