CAS RN 5786-21-0
Clozapine
CAS: 5786-21-0 Molecular Formula: C18H19ClN4 Molecular Weight (g/mol): 326.83 InChI Key: ZUXABONWMNSFBN-UHFFFAOYSA-N IUPAC Name: 6-chloro-10-(4-methylpiperazin-1-yl)-2,9-diazatricyclo[9.4.0.0³,⁸]pentadeca-1,3,5,7,10,12,14-heptaene SMILES: CN1CCN(CC1)C1=C2C=CC=CC2=NC2=CC=C(Cl)C=C2N1
CERILLIANT™ Clozapine Solution, 1.0 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™
A Certified Snap-N-Spike™ Solution for use in clinical toxicology, forensic analysis, or urine drug testing applications by LC/MS or GC/MS.
Clozapine, 97%
CAS: 5786-21-0 Molecular Formula: C18H19ClN4 Molecular Weight (g/mol): 326.828 MDL Number: MFCD00153785 InChI Key: ZUXABONWMNSFBN-UHFFFAOYSA-N Synonym: clozapine,leponex,clozapin,clozaril,fazaclo,clorazil,iprox,clozapinum,asaleptin,clozapina PubChem CID: 2818 IUPAC Name: 3-chloro-6-(4-methylpiperazin-1-yl)-5H-benzo[b][1,4]benzodiazepine SMILES: CN1CCN(CC1)C2=C3C=CC=CC3=NC4=C(N2)C=C(C=C4)Cl
Clozapine 98.0+%, TCI America™
CAS: 5786-21-0 Molecular Formula: C18H19ClN4 Molecular Weight (g/mol): 326.828 MDL Number: MFCD00153785 InChI Key: ZUXABONWMNSFBN-UHFFFAOYSA-N Synonym: clozapine,leponex,clozapin,clozaril,fazaclo,clorazil,iprox,clozapinum,asaleptin,clozapina PubChem CID: 2818 IUPAC Name: 3-chloro-6-(4-methylpiperazin-1-yl)-5H-benzo[b][1,4]benzodiazepine SMILES: CN1CCN(CC1)C2=C3C=CC=CC3=NC4=C(N2)C=C(C=C4)Cl
R&D Systems™ Clozapine
CAS: 5786-21-0 Molecular Formula: C18H19ClN4