Learn More
Lkt Laboratories CAS 34233-69-7, ≥98%, Pharmacologically inert clozapine analog that activates specialized GPCRs.
SDP
Supplier:  Lkt Laboratories C4759
Encompass_Preferred
Clozapine-N-oxide is a ligand that activates engineered GPCRs called “receptors activated solely by synthetic ligands” (RASSLs) or “designer receptors exclusively activated by designer drugs” (DREADDs); this allows for the studying of GPCR signaling control in vitro and in vivo. One example of these receptors is a mutated M3 muscarinic acetylcholine receptor (mAChR) that no longer responds to acetylcholine (ACh), only clozapine-N-oxide. Clozapine-N-oxide is otherwise pharmacologically inert.
Catalog No. NC2522152
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.