Learn More
Apexbio Technology LLC D4476 301836-43-1 200mg
SDP
Supplier:  Apexbio Technology LLC A3342200
Encompass_Preferred
D4476 (CAS 301836-43-1) is a selective cell-permeable inhibitor of casein kinase 1 (CK1) and ALK5 exhibiting IC50 values of 0 3 M for CK1 and 0 5 M for ALK5 (at 0 1 mM ATP in vitro) It likely acts via ATP-competitive inhibition of CK1 showing minimal to no significant inhibitory activity against SAPK2a/p38 PKB or SGK Treatment with D4476 inhibits CK1-mediated phosphorylation of FOXO1a (Ser322 and Ser325) and RhoB (Ser185) in H4IIE and HeLa cells respectively impacting downstream processes such as nuclear export dynamics and actin cytoskeleton regulation Additionally it induces partial p53-dependent growth arrest in HCT116 cells
Catalog No. 50-000-63238
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.