Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clozapine

Catalog No. AC464280050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464280050 5 g
AC464280010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464280050 Supplier Thermo Scientific Chemicals Supplier No. 464280050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5786-21-0 | C18H19ClN4 | 326.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5786-21-0
Molecular Formula C18H19ClN4
Molecular Weight (g/mol) 326.83
InChI Key ZUXABONWMNSFBN-UHFFFAOYSA-N
IUPAC Name 6-chloro-10-(4-methylpiperazin-1-yl)-2,9-diazatricyclo[9.4.0.0³,⁸]pentadeca-1,3,5,7,10,12,14-heptaene
SMILES CN1CCN(CC1)C1=C2C=CC=CC2=NC2=CC=C(Cl)C=C2N1

Specifications

Melting Point 182.0°C to 185.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 326.82
Chemical Name or Material Clozapine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Suspected of causing genetic defects.

Suspected of damaging fertility or the unborn child.<b
Precautionary Statement GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

EINECSNumber 227-313-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.