Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLP 04.1 VOA Kit #3, Restek™
SDP

Catalog No. 06728834 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-728-834 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-728-834 Supplier Restek Corporation Supplier No. 30460
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Contains 1mL each of these mixtures.

30006: VOA Calibration Mix 1 (ketones)

  • Acetone (67-64-1)
  • 2-Butanone (MEK) (78-93-3)
  • 2-Hexanone (591-78-6)
  • 4-Methyl-2-pentanone (MIBK) (108-10-1)

30042: 502.2 Calibration Mix 1 (gases)

  • Bromomethane (methyl bromide) (74-83-9)
  • Chloroethane (ethyl chloride) (75-00-3)
  • Chloromethane (methyl chloride) (74-87-3)
  • Dichlorodifluoromethane (CFC-12) (75-71-8)
  • Trichlorofluoromethane (CFC-11) (75-69-4)
  • Vinyl chloride (75-01-4)

30456: CLP 04.1 VOA CAL2000 MegaMix Standard

  • Benzene (71-43-2)
  • Bromodichloromethane (75-27-4)
  • Bromoform (75-25-2)
  • Carbon disulfide (75-15-0)
  • Carbon tetrachloride (56-23-5)
  • Chlorobenzene (108-90-7)
  • Chloroform (67-66-3)
  • Cyclohexane (110-82-7)
  • Dibromochloromethane (124-48-1)
  • 1,2-Dibromo-3-chloropropane (DBCP) (96-12-8)
  • 1,2-Dibromoethane (EDB) (106-93-4)
  • 1,2-Dichlorobenzene (95-50-1)
  • 1,3-Dichlorobenzene (541-73-1)
  • 1,4-Dichlorobenzene (106-46-7)
  • 1,1-Dichloroethane (75-34-3)
  • 1,2-Dichloroethane (107-06-2)
  • 1,1-Dichloroethene (75-35-4)
  • cis-1,2-Dichloroethene (156-59-2)
  • trans-1,2-Dichloroethene (156-60-5)
  • 1,2-Dichloropropane (78-87-5)
  • cis-1,3-Dichloropropene (10061-01-5)
  • trans-1,3-Dichloropropene (10061-02-6)
  • Ethylbenzene (100-41-4)
  • Isopropylbenzene (cumene) (98-82-8)
  • Methyl acetate (79-20-9)
  • Methyl tert-butyl ether (MTBE) (1634-04-4)
  • Methylcyclohexane (108-87-2)
  • Methylene chloride (dichloromethane) (75-09-2)
  • Styrene (100-42-5)
  • 1,1,2,2-Tetrachloroethane (79-34-5)
  • Tetrachloroethene (127-18-4)
  • Toluene (108-88-3)
  • 1,2,4-Trichlorobenzene (120-82-1)
  • 1,1,1-Trichloroethane (71-55-6)
  • 1,1,2-Trichloroethane (79-00-5)
  • Trichloroethene (79-01-6)
  • 1,1,2-Trichloro-1,2,2-trifluoroethane (CFC-113) (76-13-1)
  • m-Xylene (108-38-3)
  • o-Xylene (95-47-6)
  • p-Xylene (106-42-3)

Specifications

Type Multi-component Standard
Quantity 1 Kit
ShelfLife 6 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.