Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CLP 257
GREENER_CHOICE

Catalog No. p-7110794
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-421-0 10 mg
52-425-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-421-0 Supplier Tocris Bioscience™ Supplier No. 5242/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective KCC2 activator

CLP 257 is a selective KCC2 K+-Cl- cotransporter activator (EC50 = 616 nM). Exhibits selectivity for KCC2 over other KCC family members, NKCC1 and GABAA receptors, as well as 55 other receptors. Enhances Cl- transport and increases KCC2 plasma membrane expression in spinal slices with reduced KCC2 function. Alleviates hypersensitivity in rats with peripheral nerve injury.

Specifications

Chemical Name or Material (5Z)-5-[(4-Fluoro-2-hydroxyphenyl)methylene]-2-(tetrahydro-1-(2H)-pyridazinyl)-4(5H)-thiazolone
CAS 1181081-71-9
Quantity 10 mg
Target KCC2 Activators
Molecular Formula C14H14FN3O2S
Formula Weight 307.34
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.