Learn More
CLP Semivolatile Calibration Kit #2 (w/o Pesticides), Restek™
SDP
Catalog No. 06801762 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
06-801-762 1 mL
1 options
Catalog No. 06-801-762 Supplier Restek Corporation Supplier No. 31462
Compounds:
  • Contains 1 mL each of these mixtures.
  • 31007: SV Calibration Mix #1 (anilines)
  • 31008: SV Calibration Mix #2 (phenols)
  • 31009: SV Calibration Mix #3 (base neutrals)
  • 31010: SV Calibration Mix #4 (base neutrals)
  • 31011: SV Calibration Mix #5 (PAHs)
  • 31013: SV Calibration Mix #7 (dichlorobenzenes)
  • 31026: 3,3'-dichlorobenzidine
Specifications
Type Multi-component Standard
Quantity 1 mL
ShelfLife 6 months