Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CLP24 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6747G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6747G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6747G100 Supplier R&D Systems Supplier No. FAB6747G100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CLP24/TMEM204 Monoclonal specifically detects CLP24/TMEM204 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CLP24/TMEM204
Applications Flow Cytometry
Classification Monoclonal
Clone 652424
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9BSN7
Gene Alias C16orf30, C16orf30chromosome 16 open reading frame 30, Claudin-like protein 24, CLP24MGC111564, FLJ20898, TMEM204, transmembrane protein 204
Gene Symbols TMEM204
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CLP24 Accession # Q9BSN7
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79652
Test Specificity Detects human CLP24 in ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.