Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLPB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143890 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143890 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143890 Supplier Invitrogen™ Supplier No. PA5143890
  • $458.50
    $415.65 / Each of 1

    Save $42.85

    9% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293 whole cell, huamn SW620 whole cell, huamn 22RV1 whole cell, huamn THP-1 whole cell, human SGC-7901 whole cell, monkey COS-7 whole cell, rat kidney tissue, mouse heart tissue, mouse HEPA1-6 whole cell. IHC: human mammary cancer tissue, human rectal cancer tissue. ICC/IF: A549 cell. Flow: K562 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

CLPB may function as a regulatory ATPase and be related to secretion/protein trafficking process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPB
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene Clpb
Gene Accession No. Q60649, Q9H078, Q9WTT2
Gene Alias AL118244; ANKCLB; ankyrin-repeat containing bacterial clp fusion; caseinolytic mitochondrial matrix peptidase chaperone subunit B; caseinolytic peptidase B; caseinolytic peptidase B protein homolog; CLPB; ClpB caseinolytic peptidase B; ClpB caseinolytic peptidase B homolog; ClpB caseinolytic peptidase B homolog (E. coli); ClpB homolog, mitochondrial AAA ATPase chaperonin; HSP78; MEGCANN; MGCA7; Skd3; suppressor of K+ transport defect 3; suppressor of potassium transport defect 3; testicular secretory protein Li 11
Gene Symbols Clpb
Host Species Rabbit
Immunogen E.coli-derived human CLPB recombinant protein (Position: Q438-I707).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20480, 65041, 81570
Target Species Human, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.