Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLPSL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567272 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567272 Supplier Invitrogen™ Supplier No. PA567272
  • $659.00
    $548.65 / Each of 1

    Save $110.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: RGSLSPTKYNL LELKESCIRN QDCETGCCQR APDNCESHCA EKGSEGSLCQ Highest antigen sequence identity to the following orthologs - mouse 29%, rat 31%.

CLPSL1 gene ontology annotations related to this gene include digestion; lipid catabolic process; response to food.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPSL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLPSL1
Gene Accession No. A2RUU4
Gene Alias C6orf127; CLPSL1; colipase like 1; colipase-like 1; colipase-like protein 1; colipase-like protein C6orf127; dJ510O8.6; ESP32
Gene Symbols CLPSL1
Host Species Rabbit
Immunogen Recombinant Human CLPSL1. Recombinant protein control fragment (Product #RP-107910).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340204
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.