Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLPTM1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553236
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553236 Supplier Invitrogen™ Supplier No. PA553236
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNVEDFDVES KFERTVNVSV PKKTRNNGTL YAYIFLHHAG VLPWHDGKQV HLVSPLTTYM VPKPEEINLL TGESDTQQIE AEKKPTSALD EPVSHWRPRL ALNVMADNFV FDGSSLPADV HRYMKMIQLG KTVHYL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.

CLPTM1L enhances cisplatin-mediated apoptosis, when overexpressed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPTM1L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLPTM1L
Gene Accession No. Q96KA5
Gene Alias C130052I12Rik; cisplatin resistance related protein CRR9p; cisplatin resistance-related protein 9; Cleft lip and palate transmembrane protein 1-like protein; CLPTM1 like; Clptm1l; CLPTM1-like; CLPTM1-like protein; CRR9; CRR9p; RGD1307896
Gene Symbols CLPTM1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLPTM1L. Recombinant protein control fragment (Product #RP-91069).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81037
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.