Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLS-VF, MP Biomedicals™

Catalog No. MP116540402 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
90 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116540402 90 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116540402 Supplier MP Biomedicals, Inc Supplier No. 116540402
Add to Cart
Add to Cart

Cell Lysis/DNA solubilizing solution for plant tissues extractions.

CLS-VF is a cell Lysis/DNA solubilizing 90 mL solution for vegetation. For plant tissues, CLS-VF is used in conjunction with a Protein Precipitation Solution (PPS).
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Quantity 90 mL
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.