Learn More
Echelon Biosciences Research Labs Class III PI3K ELISA KIT
SDP
Supplier:  Echelon Biosciences Research Labs K3000EAKIT
Encompass
Echelons Class III PI3K ELISA Kit is a competitive (96-well) ELISA assay kit that measures Class III Phosphoinositide 3-kinases activity directly through its reaction product, PtdIns(3)P (PI(3)P). This item is non returnable
Catalog No. 50-720-443
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.