Learn More
Teknova CM Glucose Broth minus Leucine and Uracil, 1000mL, Sterile. 8 pack.
SDP
Supplier:  Teknova C82358PK
Encompass_Preferred
2.0% Cerelose Anhydrous Dextrose (Glucose),0.5% Ammonium Sulfate
Catalog No. 50-238-7809
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.