Learn More
Selleck Chemical LLC CM272 5mg
SDP
Supplier:  Selleck Chemical LLC S88125MG
Encompass_Preferred
CM-272 is a novel first-in-class dual reversible inhibitor ofG9a (GLP) andDNMTs with IC50 of 8?nM, 382 nM, 85 nM, 1200 nM, 2?nM forG9a,DNMT1,DNMT3A,DNMT3B,GLP, respectively. CM-272 prolongs survival in in vivo models of haematological malignancies by at least in part inducing immunogenic cell death. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3392
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.