Learn More
Cytiva Centramate cassette Supor PES membrane 0 65 m removal rating 0 01 m EFA medium screen
Catalog No. 5000053037 Shop All Cytiva Products
Encompass
Centramate cassette Supor PES membrane 0 65 m removal rating 0 01 m EFA medium screen
Catalog No. 50-000-53037 Supplier Cytiva Supplier No. PSM65C12P1
Centramate cassette Supor PES membrane 0 65 m removal rating 0 01 m EFA medium screen