Learn More
Selleck Chemical LLC SGC-GAK-1
SDP
Supplier:  Selleck Chemical LLC S67105MG
Encompass_Preferred
SGC-GAK-1 is a potent selective and cell-active inhibitor of cyclin G-associated kinase (GAK) with Ki value of 3 1 nM and 50-fold selectivity over nearest kinase
Catalog No. 50-000-02765
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.