Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMC Rescue™ Rappel Gloves

Catalog No. 250202
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
250202 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 250202 Supplier CMC Rescue™ Supplier No. 250202
This item is not returnable. View return policy

Armor-tan leather combined with Aramid Fiber stitching provides an extremely high level of durability. Double-layered thumb webbing protects hands when rappelling, while wear-resistant double palm guards enhance grip strength.

  • Pittard's Armor-Tan leather combined with Aramid Fiber stitching for extreme durability
  • Wear-resistant double palm guards against heat, moisture, cold, and abrasion
  • Double-layer thumb webbing protects hands when rappelling
  • Anatomically segmented second layer eases grip fatigue
  • Spandex back ventilates the gloves for enhanced comfort and flexibility
  • Attached carabiner loops allow for easy clipping onto harness gear when not in use
  • Elastic at the wrist ensures a secure fit

Specifications

Back Material Spandex
Coating Material Uncoated
Color Black, Tan
Hand Specific
Material Leather, Aramid Fiber
Product Type Extrication Gloves
Size Small
For Use With (Application) Hardward Work, Tying Knots, Setting up a System, and Rappelling
Glove Cuff Style Elasticated
Palm Type Wear-Resistant Double Palm
Series 25020
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.