Learn More
MBL International Corporation CircuLex CML/Nε-(Carboxymethyl)lysine ELISA Kit
SDP
Supplier:  MBL International Corporation CY8066
Encompass_Preferred
Application:ELISA; Size:96 Assays; Target:CML/Nε-(Carboxymethyl)lysine; The CycLex Research Product CircuLex CML/Nε-(Carboxymethyl)lysine ELISA Kit is used for the quantitative measurement of CML-adducts in mammalian serum. It can be used for 96 Assays.CircuLex CML/Nɛ-Carboxymethyl)lysine ELISA Kit.
Catalog No. 50-456-06
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.