Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CML66 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554716 Supplier Invitrogen™ Supplier No. PA554716
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKKNEGLTWP ELVIGDKQGE LIRDSAQCAA IAERLMHLTS EELNPNPDKE KPPCNAQELE ECDIFFEESS SLCRFDGNTL KTTHVVNLGS NQYL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 96%.

CML66, also known as TSGA10 (Testis Specific Gene A10), is a gene located on chromosome 8q23 that encodes a broadly immunogenic tumor antigen. The gene is expressed in various leukemias and solid tumors but has restricted expression in normal tissues, being predominantly found in the testis and heart. The physiological function of CML66 remains unclear. However, its immunogenic properties, including the ability to elicit strong humoral immune responses, make it a potential target for cancer immunotherapy. Studies have identified cytotoxic T lymphocyte (CTL) epitopes derived from CML66, suggesting that TSGA10-targeted therapies could be feasible. Moreover, research into the gene has revealed the existence of different isoforms, such as CML66-L (long) and CML66-S (short), which may have distinct roles in tumorigenesis and immune response modulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CML66
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUDCD1
Gene Accession No. Q96RS6
Gene Alias 4921532K09Rik; AA407246; AW260430; AW556235; chronic myelogenous leukemia tumor antigen 66; CML66; CML66-L; NudC domain containing 1; nudC domain-containing protein 1; Nudcd1; OVA66; RGD1310624; Tumor antigen CML66
Gene Symbols NUDCD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CML66. Recombinant protein control fragment (Product #RP-93769).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362906, 84955
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.