Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CMP-Sialic Acid, Disodium Salt, Calbiochem™,

Catalog No. 23326425MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
23-326-425MG 25 mg Glass Bottle
23-326-45MG 5 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-326-425MG Supplier MilliporeSigma™ Supplier No. 23326425MG
Add to Cart
Add to Cart

Recommended Storage

Stock solutions are Stable for up to 3 months at −20°C Unstable under acidic conditions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3063-71-6
Molecular Formula C20H31N4O16P
Molecular Weight (g/mol) 614.454
InChI Key TXCIAUNLDRJGJZ-CEUYIXQXSA-N
Synonym CMP-N-Acetylneuraminic Acid, 2Na, CMP-NANA, CMP-Neu5Ac
PubChem CID 131632558
IUPAC Name (2R,4S,5R,6R)-5-acetamido-2-[[(2R,3R,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-4-hydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
SMILES CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)OP(=O)(O)OCC2C(C(C(O2)N3C=CC(=NC3=O)N)O)O)O

Specifications

Color White
Packaging Glass Bottle
Quantity 25 mg
Percent Purity ≥95% by HPLC
Recommended Storage Stock solutions are Stable for up to 3 months at −20°C Unstable under acidic conditions.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.