Learn More
CMPK1 Mouse anti-Human, Clone: OTI1A1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505389

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Catalyzes the phosphorylation of pyrimidine nucleoside monophosphates at the expense of ATP. Plays an important role in de novo pyrimidine nucleotide biosynthesis. Has preference for UMP and CMP as phosphate acceptors. Also displays broad nucleoside diphosphate kinase activity.Specifications
CMPK1 | |
Monoclonal | |
Unconjugated | |
CMPK1 | |
CK, CMK, CMPK, UMK, UMP-CMPK, UMPK | |
Mouse | |
Affinity Chromatography | |
RUO | |
51727 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1A1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P30085 | |
CMPK1 | |
Full length human recombit protein of human CMPK1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.