Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMTM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553089
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553089 Supplier Invitrogen™ Supplier No. PA553089
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DPEHAKPESS EAPSGNLKQP ETAAALASSG SVD Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 42%.

Chemokines are members of a superfamily of small inducible, secreted, pro-inflammatory cytokines. Members of the chemokine-like factor superfamily (CKLFSF) share similarities with both chemokine and transmembrane 4 superfamilies of signaling molecules. As a member of the CKLFSF family of proteins, CMTM1 (CKLF-like MARVEL transmembrane domain-containing protein 1), also known as CKLFSF1 (Chemokine-like factor superfamily member 1), is a 169 amino acid multi-pass membrane protein that is highly expressed in testis, therefore probably playing a role in spermatogenesis or testicular development. CMTM1 contains a CC-chemokine motif and four transmembrane segments. The gene encoding CMTM1 resides in a tight gene cluster on human chromosome 16 with CKLF, CMTM2, CMTM3 and CMTM4. There are at least sixteen isoforms of CMTM1 that are produced as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMTM1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CMTM1
Gene Accession No. Q8IZ96
Gene Alias chemokine-like factor super family 1; chemokine-like factor superfamily 1; chemokine-like factor superfamily member 1; chemokine-like factor-like protein CKLFH1; CHLFH1a; CKLF like MARVEL transmembrane domain containing 1; CKLFH; CKLFH1; CKLF-like MARVEL transmembrane domain containing 1; CKLF-like MARVEL transmembrane domain-containing protein 1; CKLFSF1; CMTM1
Gene Symbols CMTM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CMTM1. Recombinant protein control fragment (Product #RP-90924).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113540
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.