Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CMTM8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103745 Supplier Invitrogen™ Supplier No. PA5103745
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CKLF8.

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMTM8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CMTM8
Gene Accession No. Q8IZV2, Q9CZR4
Gene Alias 2700018N07Rik; AA408515; chemokine-like factor super family 8; chemokine-like factor superfamily 8; chemokine-like factor superfamily member 8; chemokine-like related protein 8; CKLF like MARVEL transmembrane domain containing 8; CKLF-like MARVEL transmembrane domain containing 8; CKLF-like MARVEL transmembrane domain-containing protein 8; CKLFSF8; CKLFSF8-V2; Cmtm8; Unknown (protein for MGC:140079)
Gene Symbols CMTM8
Host Species Rabbit
Immunogen A synthesized peptide derived from human CMTM8(Accession Q8IZV2), corresponding to amino acid residues L109-N159.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152189, 70031
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.