Learn More
Enzo Life Sciences CMV cell lysate antigen (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2690100
Encompass_Preferred
Alternative name: Cytomegalovirus cell lysate antigen. Formulation: Liquid. In 0.5% Triton X-100 in PBS. Source: From cytomegalovirus, strain AD-169. Long Term Storage: -80°C. Applications: ELISA Positive control, Antibody capture.
Catalog No. 50-201-4082
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.