Learn More
Diagnostic Biosystems CMV COCKTAIL Cytomegalovirus (CMV Cocktail)
Supplier:  Diagnostic Biosystems PDM075R
Encompass_Preferred
This cocktail antibody stains infected cells giving a nuclear staining pattern with an immediate early and an early CMV antigen.
Catalog No. 50-238-5421
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.