Learn More
Invitrogen™ CNF1/CNF2 Monoclonal Antibody (JC4)

Description
MA1-40276 detects CNF1/CNF2 from E. coli samples. MA1-40276 has been successfully used in ELISA, functional assays and Western blot applications. The MA1-40276 immunogen is residues 169-191 of N-terminus of CNF1. Endotoxin <24 EU/mg.
Specifications
Specifications
| Antigen | CNF1/CNF2 |
| Applications | ELISA, Functional Assay, Western Blot |
| Classification | Monoclonal |
| Clone | JC4 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and no preservative |
| Gene Alias | Cytotoxic necrotizing factor 1; Cytotoxic necrotizing factor 2 |
| Host Species | Mouse |
| Immunogen | Residues 169-191 of N-terminus of CNF1. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.