Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNF1 Monoclonal Antibody (NG8)
GREENER_CHOICE

Catalog No. 01676394
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-394 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-394 Supplier Invitrogen™ Supplier No. MA140275
  • $859.00
    $710.40 / Each of 1

    Save $148.60

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-40275 detects CNF1 from E. coli samples. MA1-40275 has been successfully used in ELISA, functional assays and Western blot applications. The MA1-40275 immunogen is the C-terminus of CNF1. Endotoxin <24 EU/mg.

Cytotoxic necrotizing factor type 1 and 2 (CNF1 & CNF2) belong to a family of bacterial toxins that target the small GTP-binding Rho proteins that regulate the actin cytoskeleton. Members of this toxin family typically inactivate Rho; however, CNF1 and the highly related CNF2 activate Rho by deamidation. CNF1 is more frequently associated with E.coli strains that cause extraintestitinal infections in humans, particularly those of the urinary tract (such as cystitis, pyelonephritis and prostatitis). In CNF1-producing uropathogenic E. coli strains, CNF1 is chromosomally encoded and typically resides on a pathogenicity island that also contains hemolysin and P fimbria- related genes. Both CNF1 and the highly related, plasmid-encoded CNF2 are monomeric, cytoplasmic toxins of approximately 115 kDa. CNF1 can be structurally organized into three functional domains the N-terminal, central and the C-terminal domain. The latter exhibits the catalytic activity of the toxin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNF1
Applications ELISA, Functional Assay, Western Blot
Classification Monoclonal
Clone NG8
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene Alias Cytotoxic necrotizing factor 1
Host Species Mouse
Immunogen C-terminus of CNF1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.