Learn More
Description
MA3-901 detects CNG channel from human, mouse and rat samples. MA3-901 has been successfully used in Western blotting and immunohistochemistry procedures. By Western blot MA3-901 detects a band representing CNG channel. MA3-901 is specific to the a2 subunit of the CNG channel. The MA3-901 immunogen is a MBP-fusion protein of the C-terminal region of the rat a2 subunit (amino acids K581-D670).
Specifications
Specifications
| Antigen | CNGA3 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CNC 9C1 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.05% sodium azide |
| Gene | CNGA3 |
| Gene Accession No. | Q16281, Q9JJZ8 |
| Gene Alias | ACHM2; alpha subunit of cone photoreceptor CNG-channel; CCNC1; CCNCa; CCNCalpha; CNCG3; cng channel; CNG channel 1; CNG channel alpha-3; CNG1; CNG-1; CNG3; CNG-3; CNGA3; CNGalpha2; cone photoreceptor cGMP-gated channel alpha subunit; cone photoreceptor cGMP-gated channel subunit alpha; cyclic nucleotide gated channel alpha 3; cyclic nucleotide-gated cation channel alpha-3; Cyclic nucleotide-gated channel alpha-3; cyclic nucleotide-gated channel cone photoreceptor subunit alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
