Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNKSR3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988215
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8215 Supplier Bioss Supplier No. BS8030R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNKSR3 may be involved in transepithelial sodium transport. Regulates aldosterone-induced and ENaC-mediated sodium transport possibly through regulation of the ERK pathway.

Specifications

Antigen CNKSR3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CNKSR3
Gene Accession No. Q6P9H4
Gene Alias 6820402C05; BC024086; CNK homolog protein 3; CNK3; CNKSR family member 3; Cnksr3; Connector enhancer of kinase suppressor of ras 3; Connector enhancer of KSR 3; KOX14; MAGI1; Maguin-like protein; membrane associated guanylate kinase interacting protein-like 1; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase-interacting protein-like 1; Parturition-related protein 4; Prp4; ZNF21
Gene Symbols CNKSR3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CNKSR3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154043
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.