Learn More
Invitrogen™ CNOT4 Monoclonal Antibody (OTI1H2)

Description
CNOT4 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | CNOT4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1H2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CNOT4 |
| Gene Accession No. | O95628 |
| Gene Alias | CCR4-associated factor 4; CCR4-NOT transcription complex subunit 4; CCR4-NOT transcription complex subunit 4 L homeolog; CCR4-NOT transcription complex subunit 4 S homeolog; CCR4-NOT transcription complex subunit 4 S homeolog; CCR4-NOT transcription complex, subunit 4; CCR4-NOT transcription complex, subunit 4 isoform a; CLONE243; cnot4; cnot4.S; E3 ubiquitin-protein ligase CNOT4; hypothetical protein; negative regulator of transcription 4; Not4; NOT4 (negative regulator of transcription 4, yeast) homolog; Not4h; Not4hp; Potential transcriptional repressor NOT4Hp; RING-type E3 ubiquitin transferase CNOT4; transcriptional repressor NOT4Hp; XELAEV_18019807mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.