Learn More
CNOT4 Mouse anti-Human, Clone: OTI4C7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504475

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CNOT4 is a 575 amino acid protein with E3 ubiquitin ligase activity, mainly involved in protein modification and protein ubiquitination. It contains a ring type zinc finger domain of the C4C4 type, an RNA recognition motif and a bipartite nuclear localization signal.Studies show that CNOT4 is a component of the CCR4-NOT (CAF1) core complex that functions as general transcription regulation complex and has both a positive as well as a negative effect on the transcription of multiple functionally unrelated genes, thus controls gene expression. CNOT4 interacts with CNOT1 via C-terminus and with E2 ubiquitin ligases via C4C4 RING domain. Reports suggest that CNOT4 gets phosphorylated in response to DNA damage on consensus sites recognized by ATM and ATR.Specifications
CNOT4 | |
Monoclonal | |
Unconjugated | |
CNOT4 | |
CLONE243, NOT4, NOT4H | |
Mouse | |
Affinity Chromatography | |
RUO | |
4850 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4C7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95628 | |
CNOT4 | |
Human recombit protein fragment corresponding to amino acids 190-455 of human CNOT4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.