Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNPase Mouse anti-Human, Porcine, Rat, Clone: 1B9G9, Proteintech
SDP

Catalog No. 501737141 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7141 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7141 Supplier Proteintech Group Inc Supplier No. 667291IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.

Specifications

Antigen CNPase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1B9G9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CNP
Gene Accession No. P09543, P13233
Gene Alias CNP, CNP1, CNPase
Gene Symbols CNP
Host Species Mouse
Immunogen CNPase Fusion Protein Ag4227
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100511587, 1267, 25275
Target Species Human, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.