Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CNTD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109562 Supplier Invitrogen™ Supplier No. PA5109562
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVKQAENICR QATIQPRDNK RESQNWRALK QQLVNKFTLR LVSCVQLASK LSFRNKIISN ITVLNFLQAL GYLHTKEELL ESELDVLKSL NFRINLPTPL AYVETLLEVL GYNGCLVPAM.

CNTD1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNTD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CNTD1
Gene Accession No. Q8N815
Gene Alias 1700051C09Rik; 4921513J16Rik; 5430417M23Rik; CNTD; Cntd1; cyclin N-terminal domain containing 1; cyclin N-terminal domain-containing protein 1; RGD1564542
Gene Symbols CNTD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human CNTD1 (NP_001317151.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124817
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.