Learn More
Invitrogen™ CNTF Polyclonal Antibody

Description
PA1-9521 detects CNTF in human, mouse and rat samples. PA1-9521 has been successfully used in Western blot, ELISA,and immunohistochemistry procedures. By Western blot, PA1-9521 detects a 23 kDa protein representing CNTF. PA1-9521 immunogen is a synthetic peptide based on the human CNTF protein. Store at 4°C for up to 1 week, and -20°C as undiluted liquid long-term.
Specifications
Specifications
| Antigen | CNTF |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CNTF |
| Gene Accession No. | P20294, P26441, P51642 |
| Gene Alias | AI429687; Ciliary neurotrophic factor; ciliary neurotropic factor; Cntf; HCNTF; H-CNTF |
| Gene Symbols | CNTF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.