Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacharach™ CO2 Fyrite Fluid

Catalog No. 23122846
Change view
Click to view available options
For Use With (Equipment):
Bacharach Fyrite CO2 Indicator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
23-122846 Bacharach Fyrite CO2 Indicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-122846 Supplier Bacharach™ Supplier No. 105057

missing translation for 'validMainframeMsg'

For use with Bacharach Fyrite CO2 Indicator.

  • For testing carbon dioxide
  • Scale range: 0-20%, 0-60%
FYRITE CO2 refill fluid may develop a white insoluble precipitate upon aging. The precipitate does not affect fluid performance in any way, but if present, should be filtered out to prevent adherence to internal surfaces of the FYRITE. To filter, place a clean piece of thin cloth or netting loosely over the open CO2 FYRITE and slowly pour refill contents into top reservoir. Before disposing of cloth, rinse thoroughly with water. FYRITE fluid is corrosive.

Specifications

Detection Limits 0-20%, 0-60%
Components Measured Carbon Dioxide
For Use With (Equipment) Bacharach Fyrite CO2 Indicator
Type CO2 Fyrite Fluid
For Use With (Application) Testing carbon dioxide
Includes Pack of 3 bottles
Quantity 3 x 63.5 mL bottles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.