Learn More
Caron Products And Services CO2 Incubator, Reach-In, 25 cu. ft., Temp & RH (water pan)
Supplier:  Caron Products And Services 7400252
Encompass_Preferred
Carons 708L (25 ft3) Reach-In CO2 Incubator. Standard features: Intuitive touchscreen controls, Precise IR CO2 Sensor, Controlled Temperature, Elevated Humidity and Tool-less Removable Interior. Multiple options available, including Automatic Decon Cycle and gVapor Controlled Humidity. 230V, 60Hz.
Catalog No. NC2676805
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.