Learn More
Binder Inc CO2 incubator CB260UL-120V
SDP
Supplier:  Binder Inc 90400209
Encompass_Preferred
CB260UL-120V Standard Series CB - CO2 incubators with hot air sterilization and heat sterilizable CO2 sensor 9 4 cu ft 100-120 V 1 ph 50/60 Hz
Catalog No. 50-002-17516
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.