Learn More
Binder Inc CO2 incubator CBF260UL-120V
SDP
Supplier:  Binder Inc 90400217
Encompass_Preferred
CBF260UL-120V Standard Series CBF - CO2 incubators with hot air sterilization and humidity regulation 9 4 cu ft 100-120 V 1 ph 50/60 Hz
Catalog No. 50-002-17524
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.