Learn More
Binder Inc CO2 incubator CBF260UL-120V-OZ
SDP
Supplier:  Binder Inc 96400026
Encompass_Preferred
BINDER CO2 incubator CBF260UL-120V-OZ, with O2 control 0.2–20% and cell therapy compartmentalization, Series CBF - CO2 incubators with hot air sterilization and humidity regulation, 10 cu.ft, 120 V 1~ 50/60 Hz
Important characteristics:
- Contamination-safe humidification system without water pan
- Tried-and-tested anti-contamination concept with 180 °C hot air sterilization
- Hot-air sterilizable CO₂ sensor with single-beam infrared technology
- Seamless inner chamber made of stainless steel with flanges as shelf support system
- Fanless interior with Venturi CO₂ gas mixing nozzle
- Intuitive touchscreen controller
- Internal data logger, measured values can be read out in open format via USB
- Troubleshooting system with visual and acoustic alarms
- Interfaces: Ethernet, USB, zero-voltage alarm contact
- Fail-safe CO₂ system for protecting the pH of cell cultures
Catalog No. 96-400-026
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.