Learn More
Binder Inc CO2 incubator CBS170UL-120V-D
Supplier: Binder Inc 96400030
BINDER CO2 incubator CBS170UL-120V, with Access port 30 mm left, Series CB-S Solid.Line - CO2 incubators with hot air sterilization, 6 cu.ft, 120 V 1~ 50/60 Hz
Important characteristics:
- Tried-and-tested anti-contamination concept with 180 °C hot air sterilization
- Humidification system with condensation protection
- CO₂ gas mixing jet with Venturi effect
- CO₂ sensor with infrared technology
- LCD to display temperature and CO₂ along with additional information and alarms
- Internal data logger, measured values can be read out in open format via USB
- Tightly fitted inner door made of safety glass
- Seamless stainless steel inner chamber
- Perforated shelves, stainless steel
- Troubleshooting system with visual and acoustic alarms
- Interfaces: Ethernet, USB, zero-voltage alarm contact
- Standard access port 30 mm on the left
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.