Learn More
Caron Products And Services CO2 Incubator, Shaker Ready, 33 cu. ft., Temp, No RH
Supplier:  Caron Products And Services 7401332
Encompass_Preferred
Carons 3-Tier, Shaker-Ready CO2 Incubator is optimized for large-scale suspension cell culture applications using shakers. Standard features: 3 shaker-support shelves, internal outlets, intuitive touchscreen controls, IR CO2 sensor, and wide temp range (20-60C). 934L (33 ft3); 230V, 60Hz.
Catalog No. NC2676808
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.