Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CoA Synthase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588401 Supplier Invitrogen™ Supplier No. PA588401
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRPPYERPEL PTCLYVIGLT GISGSGKSSI AQRLKGLGAF VIDSDHLGHR AYAPGGPAYQ PVVEAFGTDI LHKDGIINRK VLGSRVFGNK KQLKILTDIM WPIIAKLARE EMDRAVAEGK RVCVIDAAVL LEAGWQNLVH EVWTAVIPET EAVRRIVERD GLSEAAAQSR LQSQMSGQQL VEQSHVVLST LWEPHITQRQ VEKAWALLQK RIPKTHQALD; Positive Samples: LO2, 293T, A-549, SKOV3, Mouse heart, Mouse liver, Rat kidney; Cellular Location: Cytoplasm, Mitochondrion matrix.

Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. COASY is a bifunctional enzyme that catalyzes the 2 last steps in CoA synthesis. These activities are performed by 2 separate enzymes, phosphopantetheine adenylyltransferase and dephospho-CoA kinase, in prokaryotes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CoA Synthase
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 6.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene COASY
Gene Accession No. Q13057, Q9DBL7
Gene Alias 1300003G02Rik; bifunctional coenzyme A synthase; bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase; CoA synthase; Coasy; Coenzyme A synthase; Dephospho-CoA kinase; Dephospho-CoA pyrophosphorylase; Dephosphocoenzyme A kinase; DPCK; DPCOAK; NBIA6; NBP; nucleotide binding protein; Pantetheine-phosphate adenylyltransferase; Phosphopantetheine adenylyltransferase; phosphopantetheine adenylyltransferase / dephosphocoenzyme A kinase; POV-2; PPAT; PSEC0106; Ukr1
Gene Symbols COASY
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 345-564 of human CoA synthase (COASY) (NP_0795095).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287711, 71743, 80347
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.