Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Coagulation Tubes with Sodium Citrate Solution

Catalog No. 22040046
Change view
Click to view available options
Capacity (Metric):
2 mL
2.7 mL
3.5 mL
Closure Color:
Blue with Black Ring
Blue with White Ring
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
22-040-046 3.5 mL Blue with Black Ring
22-040-045 2 mL Blue with White Ring
22-040-119 2.7 mL Blue with Black Ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-040-046 Supplier Greiner Bio-One Supplier No. 454332
Add to Cart
Add to Cart
This item is not returnable. View return policy

For routine hemostasis testing

 

  • 13 × 75mm tubes filled with buffered tri-sodium citrate solution
  • 3.2 % (0.109 mol/L) concentration
  • Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • 18-month shelf life from date of manufacture
  • Blue nonscrew safety pull cap can be removed with a gentle pull

 

TRUSTED_SUSTAINABILITY

Specifications

Product Type Sodium Citrate Blood Collection Tube
Additive 3.2% Citrate Solution
Material PET
Capacity (Metric) 3.5 mL
Closure Color Blue with Black Ring
Tube Size 13 x 75 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.